ContactUsLinkComponent
Acetate plate seal, MP Biomedicals
Acetate plate seal, MP Biomedicals
Katalog # ICNA097640105
Leverandør:  MP Biomedicals
Acetate plate seal, MP Biomedicals
Katalog # ICNA097640105
Leverandør:  MP Biomedicals
Specifikationer
  • Beskrivelse:
    Acetate plate seal, 133×83 mm sheets
  • Pakn:
    100
Specifikationer
Om denne vare
Clear adhesive seals for multiwell plates and tissue culture plates.
  • Easy to apply
  • Suitable for both short and long term storage
  • Provides tight seal to minimise evaporation and condensation