Orders LinkContactUsLinkComponent
Precision hydrometers according to Baumé
Precision hydrometers according to Baumé
Supplier:   Ludwig Schneider
Precision hydrometers according to Baumé
Supplier:  Ludwig Schneider
Certificates
Precision hydrometers according to Baumé
2 Options Available Below
Product Details & Documents
These precision hydrometers according to Baumé are suitable for calibration.
  • Without thermometer
Ordering information: Available with either a works or a DAkkS calibration certificate on request.
Packaging: In a square plastic sleeve.
Specifications for Product Family
Specifications
  • Catalog No:
  • SCNL3281222
  • SCNL3211075-20
  • Referencetemperatur:
  • 15 °C
  • 20 °C
Specifications
Product Family Options
Reference temperature: 15 °C
Product Information
  • Område
    Inddeling
    Længde
    Availability
    Price
  • 15 - 30 °Bé
    0,5 °Bé
    220 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 124,00 kr
    124,00 kr
    1 * 1 Items
       124,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    SCNL3281222
    Referencetemperatur
    15 °C
Reference temperature: 20 °C
Product Information
  • Område
    Inddeling
    Længde
    Availability
    Price
  • 40 - 50 °Bé
    0,1 °Bé
    270 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 149,00 kr
    149,00 kr
    1 * 1 Items
       149,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    SCNL3211075-20
    Referencetemperatur
    20 °C