Orders LinkContactUsLinkComponent
Test strips, nitrate/nitrite, Dosatest
Test strips, nitrate/nitrite, Dosatest
Nitrate test strips 0 - 500mg/l
Nitrite test strips 0 - 80 mg/l, 0 · 10 · 25 · 50 · 100 · 250 · 500
0 · 1 · 5 · 10 · 20 · 40 · 80, 100
Katalog # 85439.601
Leverandør:   VWR Chemicals
Test strips, nitrate/nitrite, Dosatest
Katalog # 85439.601
Leverandør:  VWR Chemicals
CAS nummer:  
Test strips, nitrate/nitrite, Dosatest
Test strips, nitrate/nitrite, Dosatest
Nitrate test strips 0 - 500mg/l
Nitrite test strips 0 - 80 mg/l, 0 · 10 · 25 · 50 · 100 · 250 · 500
0 · 1 · 5 · 10 · 20 · 40 · 80, 100
CAS nummer:  
Price
230,00 kr
Log ind for tilgængelighed
1 * 100 Items
Product Details & Documents
A ready to use kit that fits into your lab coat pocket.
  • Rapid and convenient
  • Easy to handle
Semi-quantitative results can be obtained between 10 and 120 seconds.
The colour charts are adjusted and checked using certified standard solutions that are directly traceable to primary NIST standards.
Emballering: Container with 100 test strips 6 x 95 mm.
Specifikationer
Specifikationer
  • Pakn:
    100
  • Beskrivelse:
    Nitrate test strips 0 - 500mg/l<br />Nitrite test strips 0 - 80 mg/l
  • Område:
    0 · 10 · 25 · 50 · 100 · 250 · 500<br />0 · 1 · 5 · 10 · 20 · 40 · 80
Specifikationer