Orders LinkContactUsLinkComponent
Nitric oxide assay kit
Nitric oxide assay kit
Nitric Oxide Assay Kit, 96 well, 1 KIT
Katalog # ABNOKA1641
Leverandør:   Abnova
Nitric oxide assay kit
Katalog # ABNOKA1641
Leverandør:  Abnova
CAS nummer:  
Nitric oxide assay kit
Nitric oxide assay kit
Nitric Oxide Assay Kit, 96 well, 1 KIT
CAS nummer:  
Price
4.840,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
Nitric Oxide Assay Kit for the quantitative colorimetric determination of nitric oxide in cell, food sample, plasma, serum, tissue and urine.
  • Detection range: 0,6 to 200 µm
  • Sample volume: 100 µl
Leveringsinformation: The kit contains reagent A (12 ml), reagent B (500 µl) and reagent C (12 ml), NaOH (1 ml), ZnSO4 (1 ml) and standard (1 ml).
Documents
Specifikationer
Specifikationer
  • Pakn:
    1 KIT
  • Beskrivelse:
    Nitric Oxide Assay Kit, 96 well