Solid phase extraction cartridges, CarboPrep®
CarboPrep 200 reversing cartridge, Patron, 500 mg
Katalog # REST26206
Leverandør: Restek
Gå til:
Product Details & Documents
The reversing cartridge design allows convenient inverted elution of strongly retained analytes using minimum solvent volumes. Ideal for extraction of pesticides in water.
- High adsorbent capacity (surface area ~200 m²/g) for large volume sampling
Chromatographic grade graphitised carbon provides consistent and quantitative recoveries of a wide variety of semivolatiles, pesticides and herbicides.
Specifikationer
- Kolonne form:Patron
- Beskrivelse:CarboPrep 200 reversing cartridge
- Lejevægt:500 mg