Orders LinkContactUsLinkComponent
Tin capsule kit, large
Tin capsule kit, large
Large tin capsules for most PerkinElmer 2410 applications
Katalog # PERCN2411362
Leverandør:   PerkinElmer
Tin capsule kit, large
Katalog # PERCN2411362
Leverandør:  PerkinElmer
Tin capsule kit, large
Large tin capsules for most PerkinElmer 2410 applications
Price
2.310,00 kr
Log ind for tilgængelighed
1 * 250 Items
Product Details & Documents
Large tin capsule used for the analysis of nonvolatile solid samples. It is mostly used in the combustion of the sample.
Tin capsules are routinely used to analyse polymers, soils, coal, dry dairy products, plant leaves, paper, textiles, calibration standards, and many organic and pharmaceutical products.
Specifikationer
  • Beskrivelse:
    Large tin capsules for most PerkinElmer 2410 applications