Label, clear labels
Pharma labels, transparent, 95×95 mm
Katalog # SAMP8220C-01
Leverandør: Sampling Systems
Gå til:
Product Details & Documents
These labels are made from a clear PP film. They are ideal for sealing bags and sacks.
Clear Labels are transparent so that printing underneath the label can still be easily read.
Certifications: Adhesive conforms to FDA CFR175.105.
Specifikationer
- Farve:transparent
- Beskrivelse:Pharma labels
- Dimensioner:95×95 mm
- Cat. No.:SAMP8220C-01