Orders LinkContactUsLinkComponent
Septum adapter
Septum adapter
Septum adapter for TLL syringe with septa
  HAMI31335
Septum adapter
  HAMI31335
Septum adapter
Septum adapter
Septum adapter for TLL syringe with septa
722,00 kr
1 * 1 Items
The septum adapter connects to any PTFE Luer lock (TLL) or Luer tip (LT) syringe. The adapter seals the syringe barrel but the sample remains accessible by piercing the replaceable septum.
Leveringsinformation: Supplied with 1 adapter and 12 septa.
  • Beskrivelse:
    Septum adapter for TLL syringe with septa
  • Seq:
    01