Open end wrenches
Open end wrench, 4,8 - 6,4 mm
Catalog # REST20388
Supplier: Restek
Jump to:
Product Details & Documents
High-quality miniature wrench to be used with MXT® low-dead-volume connectors.
Specifications
- Beskrivelse:Open end wrench, 4,8 - 6,4 mm