Universal cylinder wrench
REST21322
: Restek
- Beskrivelse:Universal cylinder wrench
- Pakn:1
This versatile wrench is used for tightening gauges and gas regulator CGA fittings to cylinder outlets and pipe thread connections.
This versatile wrench is used for tightening gauges and gas regulator CGA fittings to cylinder outlets and pipe thread connections.