Qualitative filter papers, grade 598, Whatman™
Supplier: Whatman products (Cytiva)
Certificates
About this item
A thick filter paper with high loading capacity. Combines medium retention with medium-fast to fast filtration speed.
These cellulose filters are used in qualitative analytical techniques to determine and identify materials. Prepleated qualitative filters are also available, which give improved flow rate and increased loading capacity compared to equivalent flat filters.
Specifications
- Catalog No:
- 515-8055
- 515-4202
- WHAT10312000
- Kvalitet:
- 598
- 598
- 598
- Vægt:
- 140 g/m²
- 140 g/m²
- 140 g/m²
- Tykkelse:
- 320 µm
- 320 µm
- 320 µm
- Partikelretention:
- 10 µm
- 10 µm
- 10 µm
- Filtreringshastighed:
- 50 s (Herzberg)
- 50 s (Herzberg)
- 50 s (Herzberg)