Orders LinkContactUsLinkComponent
Fingertight capillary column nuts
Fingertight capillary column nuts
Leverandør:   Restek
Fingertight capillary column nuts
Leverandør:  Restek
Certifikater
Fingertight capillary column nuts
Fingertight capillary column nuts
2 tilgængelige muligheder nedenfor
Product Details & Documents
These fingertight capillary column nuts rapidly tighten without wrenches and can be used with 0,25, 0,32, or 0,53 mm ID columns.
Product Family Options
Produktinformation
  • Tilgængelighed
    Price
  • REST21311
    Finger-tight capillary column nut, for use with “compact” Agilent-style ferrules
    Finger-tight capillary column nut, for use with “compact” Agilent-style ferrules
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items
  • REST21312
    Finger-tight capillary column nut, for use with standard 1,59 mm-type ferrules
    Finger-tight capillary column nut, for use with standard 1,59 mm-type ferrules