Backpressure regulator kit
Backpressure regulator kit, for Agilent GC's
Catalog # REST20634
Supplier: Restek
Jump to:
Product Details & Documents
This backpressure regulator kit increases the versatility of Agilent 5890 GC by replacing the existing 0,06 bar (207 kPa) backpressure regulator and gauge with a 4,14 bar regulator.
- Enables to use longer 60 and 105 m columns as well as shorter 10 m columns
Delivery information: Kit is supplied with complete instructions.
Specifications
- Beskrivelse:Backpressure regulator kit, for Agilent GC's