Orders LinkContactUsLinkComponent
VWR®, pH standard kit
pH standard kit, biotechnology grade, 1 KIT
Katalog # E603-KIT
Leverandør:   VWR Chemicals
VWR®, pH standard kit
Katalog # E603-KIT
Leverandør:  VWR Chemicals
CAS nummer:  
VWR®, pH standard kit
VWR®, pH standard kit
pH standard kit, biotechnology grade, 1 KIT
CAS nummer:  
Price
1.860,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
Kit contains 500 ml of pH 4,0, pH 7,0 and pH 10,0 standards.
Specifikationer
Specifikationer
  • Pakn:
    1 KIT
  • Beskrivelse:
    pH standard kit, biotechnology grade