Cleaning brushes for extractor cells
Cell cleaning brushes, Nylon, sort, Wire
Katalog # REST23999
Leverandør: Restek
Gå til:
Product Details & Documents
Firm nylon bristle brushes for easy cell cleaning and removal of solid samples.
Range of sizes to fit all extraction cells for ASE® 100, 150, 200, 300, and 350.
Specifikationer
- Børst farve:sort
- Børstemateriale:Nylon
- Handle material:Wire
- Beskrivelse:Cell cleaning brushes