Columns, MXT®-5
Om denne vare
These MXT-5 columns are Siltek-treated stainless steel with low-polarity phase, crossbond diphenyl dimethyl polysiloxane stationary phase. All residual catalysts and low molecular weight fragments are removed from the MXT-5 polymer, providing a tight monomodal distribution and extremely low bleed.
- Equivalent to USP G27, G36 phases
- 4,5 inch standard coil diameter
General purpose columns for drugs, solvent impurities, pesticides, hydrocarbons, PCB congeners, essential oils, and semivolatiles.
43 tilgængelige muligheder nedenfor
Product Details & Documents
These MXT-5 columns are Siltek-treated stainless steel with low-polarity phase, crossbond diphenyl dimethyl polysiloxane stationary phase. All residual catalysts and low molecular weight fragments are removed from the MXT-5 polymer, providing a tight monomodal distribution and extremely low bleed.
- Equivalent to USP G27, G36 phases
- 4,5 inch standard coil diameter
General purpose columns for drugs, solvent impurities, pesticides, hydrocarbons, PCB congeners, essential oils, and semivolatiles.
Product Family Options
Produktinformation
- Kolonne formFaseFilmtykkelseØ indv.LængdeTilgængelighedPrice
- REST702054,5" standard coil4,5" standard coilKapillærMXT®-50,10 µm0,25 mm15 mLog ind for tilgængelighed1 * 1 Items4.280,00 kr
- REST702084,5" standard coil4,5" standard coilKapillærMXT®-50,10 µm0,25 mm30 mLog ind for tilgængelighed1 * 1 Items7.010,00 kr
- REST718214,5" standard coil4,5" standard coilKapillærMXT®-50,20 µm0,18 mm10 mLog ind for tilgængelighed1 * 1 Items5.280,00 kr
- REST718224,5" standard coil4,5" standard coilKapillærMXT®-50,20 µm0,18 mm20 mLog ind for tilgængelighed1 * 1 Items6.990,00 kr
- REST718234,5" standard coil4,5" standard coilKapillærMXT®-50,20 µm0,18 mm40 mLog ind for tilgængelighed1 * 1 Items10.510,00 kr
- REST702204,5" standard coil4,5" standard coilKapillærMXT®-50,25 µm0,25 mm15 mLog ind for tilgængelighed1 * 1 Items4.280,00 kr
- REST702214,5" standard coil4,5" standard coilKapillærMXT®-50,25 µm0,28 mm15 mLog ind for tilgængelighed1 * 1 Items4.350,00 kr
- REST702224,5" standard coil4,5" standard coilKapillærMXT®-50,25 µm0,53 mm15 mLog ind for tilgængelighed1 * 1 Items4.470,00 kr
- REST702234,5" standard coil4,5" standard coilKapillærMXT®-50,25 µm0,25 mm30 mLog ind for tilgængelighed1 * 1 Items6.940,00 kr
- REST702244,5" standard coil4,5" standard coilKapillærMXT®-50,25 µm0,28 mm30 mLog ind for tilgængelighed1 * 1 Items7.070,00 kr
- REST702254,5" standard coil4,5" standard coilKapillærMXT®-50,25 µm0,53 mm30 mLog ind for tilgængelighed1 * 1 Items7.730,00 kr
- REST702264,5" standard coil4,5" standard coilKapillærMXT®-50,25 µm0,25 mm60 mLog ind for tilgængelighed1 * 1 Items11.420,00 kr
- REST702274,5" standard coil4,5" standard coilKapillærMXT®-50,25 µm0,28 mm60 mLog ind for tilgængelighed1 * 1 Items11.900,00 kr
- REST702284,5" standard coil4,5" standard coilKapillærMXT®-50,25 µm0,53 mm60 mLog ind for tilgængelighed1 * 1 Items12.220,00 kr
- REST718244,5" standard coil4,5" standard coilKapillærMXT®-50,40 µm0,18 mm10 mLog ind for tilgængelighed1 * 1 Items5.280,00 kr
- REST718254,5" standard coil4,5" standard coilKapillærMXT®-50,40 µm0,18 mm20 mLog ind for tilgængelighed1 * 1 Items6.990,00 kr
- REST702354,5" standard coil4,5" standard coilKapillærMXT®-50,50 µm0,25 mm15 mLog ind for tilgængelighed1 * 1 Items4.240,00 kr
- REST702364,5" standard coil
- REST702374,5" standard coil4,5" standard coilKapillærMXT®-50,50 µm0,53 mm15 mLog ind for tilgængelighed1 * 1 Items4.470,00 kr
- REST702384,5" standard coil4,5" standard coilKapillærMXT®-50,50 µm0,25 mm30 mLog ind for tilgængelighed1 * 1 Items6.940,00 kr
- REST702394,5" standard coil4,5" standard coilKapillærMXT®-50,50 µm0,28 mm30 mLog ind for tilgængelighed1 * 1 Items7.070,00 kr
- REST702404,5" standard coil4,5" standard coilKapillærMXT®-50,50 µm0,53 mm30 mLog ind for tilgængelighed1 * 1 Items7.730,00 kr
- REST702414,5" standard coil
- REST702504,5" standard coil4,5" standard coilKapillærMXT®-51,00 µm0,25 mm15 mLog ind for tilgængelighed1 * 1 Items4.240,00 kr
- REST702514,5" standard coil4,5" standard coilKapillærMXT®-51,00 µm0,28 mm15 mLog ind for tilgængelighed1 * 1 Items4.350,00 kr
- REST70252-2733,5" coil
- REST702524,5" standard coil4,5" standard coilKapillærMXT®-51,00 µm0,53 mm15 mLog ind for tilgængelighed1 * 1 Items4.470,00 kr
- REST702534,5" standard coil4,5" standard coilKapillærMXT®-51,00 µm0,25 mm30 mLog ind for tilgængelighed1 * 1 Items6.940,00 kr
- REST702544,5" standard coil4,5" standard coilKapillærMXT®-51,00 µm0,28 mm30 mLog ind for tilgængelighed1 * 1 Items7.070,00 kr
- REST70255-2733,5" coil
- REST702554,5" standard coil4,5" standard coilKapillærMXT®-51,00 µm0,53 mm30 mLog ind for tilgængelighed1 * 1 Items7.730,00 kr
- REST702574,5" standard coil4,5" standard coilKapillærMXT®-51,00 µm0,28 mm60 mLog ind for tilgængelighed1 * 1 Items11.900,00 kr
- REST702584,5" standard coil4,5" standard coilKapillærMXT®-51,00 µm0,53 mm60 mLog ind for tilgængelighed1 * 1 Items12.220,00 kr
- REST702674,5" standard coil4,5" standard coilKapillærMXT®-51,50 µm0,53 mm15 mLog ind for tilgængelighed1 * 1 Items4.470,00 kr
- REST702704,5" standard coil4,5" standard coilKapillærMXT®-51,50 µm0,53 mm30 mLog ind for tilgængelighed1 * 1 Items7.730,00 kr
- REST702814,5" standard coil
- REST702824,5" standard coil4,5" standard coilKapillærMXT®-53,00 µm0,53 mm15 mLog ind for tilgængelighed1 * 1 Items4.470,00 kr
- REST702844,5" standard coil4,5" standard coilKapillærMXT®-53,00 µm0,28 mm30 mLog ind for tilgængelighed1 * 1 Items7.070,00 kr
- REST702854,5" standard coil4,5" standard coilKapillærMXT®-53,00 µm0,53 mm30 mLog ind for tilgængelighed1 * 1 Items7.730,00 kr
- REST702884,5" standard coil4,5" standard coilKapillærMXT®-53,00 µm0,53 mm60 mLog ind for tilgængelighed1 * 1 Items12.220,00 kr
- REST702774,5" standard coil4,5" standard coilKapillærMXT®-55,00 µm0,53 mm15 mLog ind for tilgængelighed1 * 1 Items4.600,00 kr
- REST702794,5" standard coil4,5" standard coilKapillærMXT®-55,00 µm0,53 mm30 mLog ind for tilgængelighed1 * 1 Items7.790,00 kr
- REST702834,5" standard coil4,5" standard coilKapillærMXT®-55,00 µm0,53 mm60 mLog ind for tilgængelighed1 * 1 Items12.220,00 kr