Orders LinkContactUsLinkComponent
Capillary columns, MXT®-502.2
Capillary columns, MXT®-502.2
Supplier:   Restek
Capillary columns, MXT®-502.2
Supplier:  Restek
Certificates
Capillary columns, MXT®-502.2
Capillary columns, MXT®-502.2
4 Options Available Below
Product Details & Documents
These Siltek®-treated stainless steel MXT®-502.2 columns enable to quantify all compounds when using a mass spectrometer or a PID in tandem with an ELCD. A 105 m column can separate the light gases specified in EPA methods without subambient cooling.
  • Proprietary Crossbond
  • 114,3 mm standard coil diameter
  • Diphenyl/dimethyl polysiloxane phase provides low bleed and thermal stability to 320 °C
These application specific columns with unique selectivity are ideal for volatile organic pollutants and in many gasoline range organics (GRO) methods for monitoring underground storage tanks. Separation of trihalomethanes and ideal polarity for light hydrocarbons and aromatics.
Product Family Options
Product Information
  • Kolonne form
    Fase
    Filmtykkelse
    Ø indv.
    Længde
    Availability
    Price
  • Kapillær
    MXT®-502.2
    1,60 µm
    0,28 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 7.380,00 kr
    7.380,00 kr
    1 * 1 Items
       7.380,00 kr
  • Kapillær
    MXT®-502.2
    3,00 µm
    0,53 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 8.170,00 kr
    8.170,00 kr
    1 * 1 Items
       8.170,00 kr
  • Kapillær
    MXT®-502.2
    3,00 µm
    0,53 mm
    60 m
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
  • Kapillær
    MXT®-502.2
    3,00 µm
    0,53 mm
    105 m
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 15.670,00 kr
    15.670,00 kr
    1 * 1 Items
       15.670,00 kr