Density hydrometers
Leverandør: Ludwig Schneider
Om denne vare
Precision hydrometers with integrated blue filling thermometer.
- Suitable for calibration
Error limit: ±0,5 kg/m³
Reference temperature: 15 °C
: ASTM E2995-14
: A works or DAkkS calibration certificate is available on request.
: Each packed in a square plastic sleeve.
12 tilgængelige muligheder nedenfor
Product Details & Documents
Precision hydrometers with integrated blue filling thermometer.
- Suitable for calibration
Error limit: ±0,5 kg/m³
Reference temperature: 15 °C
: ASTM E2995-14
: A works or DAkkS calibration certificate is available on request.
: Each packed in a square plastic sleeve.
Specifications for Product Family
Specifikationer
- Catalog No:
- SCNL3222111S
- SCNL3222112S
- SCNL3222113S
- SCNL3222114S/04/3P
- SCNL3222114S
- SCNL3222115S
- SCNL3222116S/04/3P
- SCNL3222116S
- SCNL3222117S
- SCNL3222118S
- SCNL3222119S
- SCNL3222120S
- Referencetemperatur:
- 15 °C
- 15 °C
- 15 °C
- 15 °C
- 15 °C
- 15 °C
- 15 °C
- 15 °C
- 15 °C
- 15 °C
- 15 °C
- 15 °C
Specifikationer
Product Family Options
Produktinformation
- ModelOmrådeInddelingTemperaturområdeLængdeTilgængelighedPrice
- S500 HL-140,600 - 0,650 g/ml0,0005 g/ml−20...+65 °C in 1 °C380 mmLog ind for tilgængelighed1 * 1 Items658,00 krSpecifikationer:
More
Specifications
Cat. No.SCNL3222111SReferencetemperatur15 °C - S501 HL-140,650 - 0,700 g/ml0,0005 g/ml−20...+65 °C in 1 °C380 mmLog ind for tilgængelighed1 * 1 Items658,00 krSpecifikationer:
More
Specifications
Cat. No.SCNL3222112SReferencetemperatur15 °C - S502 HL-140,700 - 0,750 g/ml0,0005 g/ml−20...+65 °C in 1 °C380 mmLog ind for tilgængelighed1 * 1 Items614,00 krSpecifikationer:
More
Specifications
Cat. No.SCNL3222113SReferencetemperatur15 °C - S503 HL-140,750 - 0,800 g/ml0,0005 g/ml−20...+65 °C in 1 °C380 mmLog ind for tilgængelighed1 * 1 Items4.480,00 krSpecifikationer:
More
Specifications
Cat. No.SCNL3222114S/04/3PReferencetemperatur15 °C - S503 HL-140,750 - 0,800 g/ml0,0005 g/ml−20...+65 °C in 1 °C380 mmLog ind for tilgængelighed1 * 1 Items614,00 krSpecifikationer:
More
Specifications
Cat. No.SCNL3222114SReferencetemperatur15 °C - S504 HL-140,800 - 0,850 g/ml0,0005 g/ml−20...+65 °C in 1 °C380 mmLog ind for tilgængelighed1 * 1 Items614,00 krSpecifikationer:
More
Specifications
Cat. No.SCNL3222115SReferencetemperatur15 °C - S505 HL-140,850 - 0,900 g/ml0,0005 g/ml−20...+65 °C in 1 °C380 mmLog ind for tilgængelighed1 * 1 Items4.480,00 krSpecifikationer:
More
Specifications
Cat. No.SCNL3222116S/04/3PReferencetemperatur15 °C - S505 HL-140,850 - 0,900 g/ml0,0005 g/ml−20...+65 °C in 1 °C380 mmLog ind for tilgængelighed1 * 1 Items614,00 krSpecifikationer:
More
Specifications
Cat. No.SCNL3222116SReferencetemperatur15 °C - S506 HL-140,900 - 0,950 g/ml0,0005 g/ml−20...+65 °C in 1 °C380 mmLog ind for tilgængelighed1 * 1 Items614,00 krSpecifikationer:
More
Specifications
Cat. No.SCNL3222117SReferencetemperatur15 °C - S507 HL-140,950 - 1,000 g/ml0,0005 g/ml−20...+65 °C in 1 °C380 mmLog ind for tilgængelighed1 * 1 Items614,00 krSpecifikationer:
More
Specifications
Cat. No.SCNL3222118SReferencetemperatur15 °C - S508 HL-141,000 - 1,050 g/ml0,0005 g/ml−20...+65 °C in 1 °C380 mmLog ind for tilgængelighed1 * 1 Items658,00 krSpecifikationer:
More
Specifications
Cat. No.SCNL3222119SReferencetemperatur15 °C - S509 HL-141,050 - 1,100 g/ml0,0005 g/ml−20...+65 °C in 1 °C380 mmLog ind for tilgængelighed1 * 1 Items658,00 krSpecifikationer:
More
Specifications
Cat. No.SCNL3222120SReferencetemperatur15 °C