Orders LinkContactUsLinkComponent
Diatomaceous earth sorbents
Diatomaceous earth, bulk adsorbent, in 1 kg packs, 1
Katalog # REST26033
Leverandør:   Restek
Diatomaceous earth sorbents
Katalog # REST26033
Leverandør:  Restek
CAS nummer:  
Diatomaceous earth sorbents
Diatomaceous earth sorbents
Diatomaceous earth, bulk adsorbent, in 1 kg packs, 1
CAS nummer:  
Price
2.740,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Diatomaceous earth is used as a filter aid to improve extraction efficiency of densely packed soils, such as clays. By mixing the sample with diatomaceous earth, recoveries can be improved and excess moisture can be absorbed.
  • Improves extraction efficiency
  • Adsorbs moisture from samples
Specifikationer
Specifikationer
  • Pakn:
    1
  • Beskrivelse:
    Diatomaceous earth, bulk adsorbent, in 1 kg packs
Specifikationer