Orders LinkContactUsLinkComponent
Safety caps, DIN 50 threads
Safety caps, DIN 50 threads
Safety caps, DIN 50 threads
Safety caps, DIN 50 threads
DIN50 safety caps seal to the bottle/canister with an O-ring behind a PTFE lip, so the O-ring is therefore not a wetted part. Unused ports can be closed with the stopcock.
  • Built-in stopcocks for easy interruption of solvent lines
  • Fit all bottles or canisters with DIN50 thread
  • Prevent evaporation of volatile compounds
  • Stopcocks rated up to 4,96 bar gas at 25 °C
Ordering information: A bottom-sealing barbed adapter to connect 1/8" (3,2 mm) ID tubing can be ordered separately.
Product Family Options
  • Beskrivelse
    Gevind
    Stik
    Materiale
  • Safety cap, with stopcock, O-ring, nuts, ferrules and colour-sleeve adapters
    DIN50
    Two ports with 1/4"-28 thread for 1/8" or 1/16" O.Ø tubing
    PP
  • Safety cap, with stopcock, O-ring, nuts, ferrules and colour-sleeve adapters
    DIN50
    Three ports with 1/4"-28 thread for 1/8" or 1/16" O.Ø tubing
    PP
    1 * 1 Items
    1 * 1 Items - 3.260,00 kr
    3.260,00 kr
    1 * 1 Items
       3.260,00 kr