pH software, EasyDirect™
License key for EasyDirect™ pH software
Katalog # 661-0271
Leverandør: Mettler - Toledo
Gå til:
Product Details & Documents
This intuitive software for improved data management can be used with SevenCompact™, SevenCompact™ Duo, Seven2Go™ Pro, FiveEasyPlus or SevenExcellence™ pH meters. Within a few clicks the data measured on the pH meter can be seamlessly transferred to a computer. The application will store the data unmodified and allow for further printing or analysis.
- Available in 12 languages
- Data can be stored, printed or exported to Excel or any other csv application
- Up to three instruments can be connected simultaneously
Bestillingsoplysninger: Supplied without meter, computer and USB cable.
Specifikationer
- Beskrivelse:License key for EasyDirect™ pH software
- Seq:40
Accessories
Produktinformation
- Katalog #TilgængelighedPrice
- USB-kabel 1,8 m til pc eller printer