Labels for BMP™ 51/53 printer, type B-427
Self-laminating labels, B-427, Vinyl, Permanent akryl, Sort på hvid, BMP41, 27×6 mm, 240 labels/box
Katalog # BRDY131575
Leverandør: Brady
Gå til:
Product Details & Documents
These self-laminating vinyl labels are ideal for labelling highly curved surfaces, wires and cables. The labels feature a white printable area with clear 'tail' that wraps around marker that serves as over-laminate to protect the text, providing superior resistance to abrasion, dirt, oils, solvents, water and excessive handling.
Specifikationer
- Printer:BMP41
- Model:B-427
- Farve:Sort på hvid
- Beskrivelse:Self-laminating labels
- Dimensioner:27×6 mm
- Materiale:Vinyl
- Seq:18
- Pakket:240 labels/box
- Klæbestof:Permanent akryl