Orders LinkContactUsLinkComponent
Labels for BMP™ 71 printer, type B-499
Printer labels, B-499, Nylon, Permanent akryl, hvid, BMP71, 12×38 mm, 500 mærkater/rulle
Katalog # BRDY114802
Leverandør:   Brady
Labels for BMP™ 71 printer, type B-499
Katalog # BRDY114802
Leverandør:  Brady
Labels for BMP™ 71 printer, type B-499
Labels for BMP™ 71 printer, type B-499
Printer labels, B-499, Nylon, Permanent akryl, hvid, BMP71, 12×38 mm, 500 mærkater/rulle
Price
Spørg for pris
Log ind for tilgængelighed
1 * 1 ROL
Product Details & Documents
Specifikationer
  • Printer:
    BMP71
  • Model:
    B-499
  • Farve:
    hvid
  • Beskrivelse:
    Printer labels
  • Dimensioner:
    12×38 mm
  • Materiale:
    Nylon
  • Seq:
    06
  • Pakket:
    500 mærkater/rulle
  • Klæbestof:
    Permanent akryl