Orders LinkContactUsLinkComponent
Conical bend testers
Conical bend testers
Leverandør:   Industrial Physics
Conical bend testers
Leverandør:  Industrial Physics
Certifikater
Conical bend testers
Conical bend testers
2 tilgængelige muligheder nedenfor
Product Details & Documents
These Bend Test Mandrels are designed to bend coated panels over a conical shaped mandrel in order to assess the elasticity or resistance of a coating, paint or varnish to cracking, elongation and/or detachment from a metal test panel.
  • Sturdy apparatus in a combination of anodized aluminium and stainless steel
  • Large knob on bending arm for easy and smooth bending
  • Quick lock panel clamp (for Pro model only)
The 'Pro' model is secured by a quick lever handle, that locks and unlocks the panel, in a split second using just hand.
The 'Basic' model is secured by two clamping knobs, which have to be tightened and untightened, by turning respectively clockwise or anticlockwise.
Test panel size: 100×180 mm - Maximum panel thickness: 0,8 mm
Certifications: ISO 6860 and ASTM D522.
Documents
Product Family Options
Produktinformation
  • Model
    Tilgængelighed
    Price
  • SIMESP1830
    Conical bend test mandrel (Ø = 3,1 - 38 mm)
    Conical bend test mandrel (Ø = 3,1 - 38 mm)
    "Pro"
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 12.820,00 kr
    12.820,00 kr
    1 * 1 Items
       12.820,00 kr
  • SIMESP1831
    Conical bend test mandrel (Ø = 3,1 = 38 mm)
    Conical bend test mandrel (Ø = 3,1 = 38 mm)
    "Basic"
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 8.040,00 kr
    8.040,00 kr
    1 * 1 Items
       8.040,00 kr