Orders LinkContactUsLinkComponent
Reagensglasholdere
Reagensglasholdere
250 mm, 10 - 25 mm
Catalog # BOCH2283
Supplier:   BOCHEM
Reagensglasholdere
Catalog # BOCH2283
Supplier:  BOCHEM
Reagensglasholdere
Reagensglasholdere
250 mm, 10 - 25 mm
Price
100,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
This test tube holder is made from stainless steel (18/10) and features a wooden handle. For tubes with a diameter from 10 to 25 mm.
Specifications
  • Længde:
    250 mm
  • Dækket område:
    10 - 25 mm