Orders LinkContactUsLinkComponent
Oversized breaker lockout device
Oversized breaker lockout device
Leverandør:   Brady
Oversized breaker lockout device
Leverandør:  Brady
Certifikater
Oversized breaker lockout device
Oversized breaker lockout device
2 tilgængelige muligheder nedenfor
Product Details & Documents
This oversized 480/600 V frame lockout fits breakers up to 57,15 mm wide and 22,2 mm thick.
  • Made of durable PP and glass filled impact-modified nylon
  • Applies in seconds with thumbscrew mechanism so no screwdriver is needed
  • New blade design with improved grip works on even more breaker models
Product Family Options
Produktinformation
  • Model
    Farve
    Tilgængelighed
    Price
  • Oversized breaker lockout, 480 - 600 Volt
    sort/rød
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 204,00 kr
    204,00 kr
    1 * 1 Items
       204,00 kr
  • Oversized breaker lockout, 480 - 600 Volt
    sort/rød
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 6 Items
    1 * 6 Items - 1.010,00 kr
    1.010,00 kr
    1 * 6 Items
       1.010,00 kr