Mop handle, aluminium
Aluminium telescopic handle (blue) with tilting lever, 1,0 - 1,8 m
Katalog # PPSA2016050
Leverandør: PFENNIG REINIGUNGSTECHNIK
Gå til:
Product Details & Documents
Aluminium telescopic handle for mop frames.
- Height adjustable
- Weight 460 g
Documents
PPSA2016050_TDS_EN
Datablad
Specifikationer
- Beskrivelse:Aluminium telescopic handle (blue) with tilting lever
- Længde:1,0 - 1,8 m