Orders LinkContactUsLinkComponent
Bunded spill flooring
 :   asecos
Bunded spill flooring
 :  asecos
Bunded spill flooring
Bunded spill flooring
The bunded spill flooring made from galavanised steel is designed for covering a large storage area.
  • Removable, hot-dip galvanized grids
  • Optimal adaption on space demands
  • Approved for the safe storage of water pollutants and flammable liquids
Load capacity: 5 000 kg/m²
Product Family Options
  • Beskrivelse
    B×D×H
  • Bunded spill flooring, 20 l, wheel load 450 kg
    500×500×125 mm
    1 * 1 Items
    1 * 1 Items - 1.980,00 kr
    1.980,00 kr
    1 * 1 Items
       1.980,00 kr
  • Bunded spill flooring, 40 l, wheel load 450 kg
    1000×500×125 mm
    1 * 1 Items
    1 * 1 Items - 2.430,00 kr
    2.430,00 kr
    1 * 1 Items
       2.430,00 kr
  • Bunded spill flooring, 80 l, wheel load 450 kg
    1000×1000×125 mm
    1 * 1 Items
    1 * 1 Items - 4.330,00 kr
    4.330,00 kr
    1 * 1 Items
       4.330,00 kr
  • Bunded spill flooring, 165 l, wheel load 450 kg
    2000×1000×125 mm
    1 * 1 Items
    1 * 1 Items - 7.770,00 kr
    7.770,00 kr
    1 * 1 Items
       7.770,00 kr
  • Bunded spill flooring, 95 l, wheel load 450 kg
    2500×500×125 mm
    1 * 1 Items
    1 * 1 Items - 4.780,00 kr
    4.780,00 kr
    1 * 1 Items
       4.780,00 kr
  • Bunded spill flooring, 210 l, wheel load 450 kg
    2500×1000×125 mm
    1 * 1 Items
    1 * 1 Items - 8.780,00 kr
    8.780,00 kr
    1 * 1 Items
       8.780,00 kr
Accessories
  •  
    Beskrivelse
  • MSGA8819
    Joining element, 2470×45 mm
    1 * 1 Items - 520,00 kr
  •  
    Beskrivelse
  • MSGA8820
    Ramp, 450×1120 mm
    1 * 1 Items - 2.250,00 kr
  •  
    Beskrivelse
  • MSGA8821
    Ramp, 950×1120 mm
    1 * 1 Items - 4.060,00 kr
  •  
    Beskrivelse
  • MSGA2595
    Cross joiner, 130×130 mm
    1 * 1 Items - 436,00 kr
  •  
    Beskrivelse
  • MSGA8816
    Joining element, 470×45 mm
    1 * 1 Items - 257,00 kr
  •  
    Beskrivelse
  • MSGA8817
    Joining element, 970×45 mm
    1 * 1 Items - 348,00 kr
  •  
    Beskrivelse
  • MSGA8818
    Joining element, 1970×45 mm
    1 * 1 Items - 421,00 kr
  •  
    Beskrivelse
  • MSGA8822
    Ramp, 1950×1120 mm
    1 * 1 Items - 7.230,00 kr
  •  
    Beskrivelse
  • MSGA8823
    Ramp, 2450×1120 mm
    1 * 1 Items - 8.060,00 kr
  •  
    Beskrivelse
  • MSGA8824
    Distance fastener, 25×120 mm
    1 * 1 Items - 291,00 kr
  •  
    Beskrivelse
  • MSGA8825
    Edge fastener, 20×100 mm
    1 * 1 Items - 283,00 kr
  •  
    Beskrivelse
  • MSGA8826
    Corner ramp, 1120×1120 mm
    1 * 1 Items - 4.820,00 kr