Orders LinkContactUsLinkComponent
Wedge for measuring reverse-side tolerance
Wedge for measuring reverse-side tolerance
Wedge for measuring reverse-side tolerance, range 1 - 10 mm
Katalog # SIMESP6000
Leverandør:   Industrial Physics
Wedge for measuring reverse-side tolerance
Katalog # SIMESP6000
Leverandør:  Industrial Physics
Wedge for measuring reverse-side tolerance
Wedge for measuring reverse-side tolerance
Wedge for measuring reverse-side tolerance, range 1 - 10 mm
Price
552,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Anodised aluminum wedge for quick measurement of reverse-side tolerance of windows and doors.
  • Tolerance is given in millimetres
  • Also equipped with a measuring scale for optically determining the thickness or glass
Documents
Specifikationer
  • Beskrivelse:
    Wedge for measuring reverse-side tolerance, range 1 - 10 mm