Impact test according to CSA
Impact tester according to Canadian Standards Association (CSA)
Katalog # SIMESP1896
Leverandør: Industrial Physics
Gå til:
Product Details & Documents
Impact test used to determine the impact resistivity and flexibility of materials.
- Spirit level built-in for correct positioning
- Dual scale instrument equipped with a special guidance
- Distance between each impact is always according to the standards
Certifications: Conforms to the Canadian Standards Association (CSA).
Documents
TQC Company Video
Videoer
Specifikationer
- Beskrivelse:Impact tester according to Canadian Standards Association (CSA)