Orders LinkContactUsLinkComponent
Water testing kit, MINIKIT
Leverandør:   Lovibond
Water testing kit, MINIKIT
Leverandør:  Lovibond
Certifikater
Water testing kit, MINIKIT
Water testing kit, MINIKIT
19 tilgængelige muligheder nedenfor
Product Details & Documents
MINIKIT provides quick and simple tests for the analysis of boiler water, cooling water, drinking and industrial water.
  • MINIKITS are based on titrimetric methods
  • Easy operation and exact reagent dosing
  • Foil-wrapped tablet reagents with a minimum guaranteed shelf life of five years
Tablet count method: In this method the liquid titration solution and indicator are replaced by AQUALYTIC® tablet reagents. A specific number of tablets is added to a defined sample volume until a chemically induced colour change takes place. The concentration of the parameter being measured is calculated from the number of tablets required. The measuring range can be expanded by varying the sample volume.
Yes/No test: This method tells whether a specific ingredient is present in the water and/or if its concentration is higher or lower than a defined level.
Turbidity method: A two-section calibrated test tube is filled with the water sample and a reagent tablet added. The reagent creates a level of turbidity that is proportional to the concentration of the parameter being measured. The inner tube, which has a black dot on its base, is lowered until the dot is obscured by the turbidity. The result is read off from the water level in the inner tube.
Product Family Options
Tablet count method
Produktinformation
  • Område
    Model
    Pakn
    Tilgængelighed
    Price
  • LOVI414100
    Cleaning Acid Strength
    Cleaning Acid Strength
    0,75 - 10 % acid
    AF 410
    1 KIT
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 719,00 kr
    719,00 kr
    1 * 1 Each
       719,00 kr
  • LOVI414110
    Organo-Phosphonate
    Organo-Phosphonate
    1 - 20 mg/l active O-P
    AF 411
    1 KIT
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 2.100,00 kr
    2.100,00 kr
    1 * 1 Each
       2.100,00 kr
  • 705-0410
    Alkalinity, Total (M)
    Alkalinity, Total (M)
    10 - 500 mg/l CaCO3, ~0,1 - 5 mmol/l
    AF 413
    1 KIT
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 719,00 kr
    719,00 kr
    1 * 1 Each
       719,00 kr
  • 705-0411
    Alkalinity, P
    Alkalinity, P
    20 - 500 mg/l CaCO3
    AF 414
    1 KIT
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 719,00 kr
    719,00 kr
    1 * 1 Each
       719,00 kr
  • 705-0413
    Alkalinity Caustic/P
    Alkalinity Caustic/P
    20 - 500 mg/l CaCO3
    AF 415
    1 KIT
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 719,00 kr
    719,00 kr
    1 * 1 Each
       719,00 kr
  • 705-0414
    Calcium Hardness
    Calcium Hardness
    10 - 500 mg/l CaCO3, ~ 0,1 - 5 mmol/l
    AF 416
    1 KIT
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 719,00 kr
    719,00 kr
    1 * 1 Each
       719,00 kr
  • Chloride
    5 - 5000 mg/l Cl
    AF 418
    1 KIT
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 955,00 kr
    955,00 kr
    1 * 1 Each
       955,00 kr
  • Hardness Total
    5 - 500 mg/l CaCO3, ~ 0,05 - 5 mmol/l
    AF 424
    1 KIT
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 698,00 kr
    698,00 kr
    1 * 1 Each
       698,00 kr
  • LOVI414250
    Hardness Total (low range)
    Hardness Total (low range)
    1 - 50 mg/l CaCO3, ~ 0,01 - 0,5 mmol/l
    AF 425
    1 KIT
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 719,00 kr
    719,00 kr
    1 * 1 Each
       719,00 kr
  • LOVI414260
    Hardness Total (very low range)
    Hardness Total (very low range)
    1 - 10 mg/l CaCO3, ~ 0,01 - 0,1 mmol/l
    AF 426
    1 KIT
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 719,00 kr
    719,00 kr
    1 * 1 Each
       719,00 kr
  • 705-0417
    Nitrite
    Nitrite
    70 - 1500 mg/l NaNO2
    AF 427
    1 KIT
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 719,00 kr
    719,00 kr
    1 * 1 Each
       719,00 kr
  • LOVI414320
    Sulphate (low range)
    Sulphate (low range)
    20 - 200 mg/l Na2SO4
    AF 432
    1 KIT
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 984,00 kr
    984,00 kr
    1 * 1 Each
       984,00 kr
  • LOVI414340
    Sulphite (low range)
    Sulphite (low range)
    2 - 50 mg/l Na2SO3
    AF 434
    1 KIT
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 719,00 kr
    719,00 kr
    1 * 1 Each
       719,00 kr
  • 705-0421
    Sulphite (high range)
    Sulphite (high range)
    20 - 500 mg/l Na2SO3
    AF 435
    1 KIT
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 719,00 kr
    719,00 kr
    1 * 1 Each
       719,00 kr
  • 705-0422
    Tannin Index
    Tannin Index
    2 - 20 units
    AF 436
    1 KIT
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 719,00 kr
    719,00 kr
    1 * 1 Each
       719,00 kr
Yes/No test method
Produktinformation
  • Område
    Model
    Pakn
    Tilgængelighed
    Price
  • 705-0420
    QAC (Quaternary Ammonium Comp.)
    QAC (Quaternary Ammonium Comp.)
    0 - 500 mg/l active QAC Limit 200 mg/l (Yes/No)
    AF 417
    1 KIT
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 911,00 kr
    911,00 kr
    1 * 1 Each
       911,00 kr
  • LOVI414230
    Hardness Total (Yes/No)
    Hardness Total (Yes/No)
    Limit 4 mg/l, 8 mg/l or 20 mg/l CaCO3, ~ 0,04 or 0,08 or 0,2 mmol/l
    AF 423
    1 KIT
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 719,00 kr
    719,00 kr
    1 * 1 Each
       719,00 kr
Turbidity test method
Produktinformation
  • Område
    Model
    Pakn
    Tilgængelighed
    Price
  • LOVI414220
    Cyanuric Acid
    Cyanuric Acid
    20 - 200 mg/l
    AF 422
    1 KIT
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 896,00 kr
    896,00 kr
    1 * 1 Each
       896,00 kr
  • LOVI414310
    Sulphate
    Sulphate
    40 - 200 mg/l SO4 (40 - 4000 mg/l by dilution)
    AF 431
    1 KIT
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 984,00 kr
    984,00 kr
    1 * 1 Each
       984,00 kr
Accessories
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Dilution/shaker tube with lid, plastic, graduated, 100 ml
    Dilution/shaker tube with lid, plastic, graduated, 100 ml
    LOVI385130
    Log ind for tilgængelighed
    1 * 1 Items - 112,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Dilution/shaker tube with lid, plastic, graduated, 30 ml
    Dilution/shaker tube with lid, plastic, graduated, 30 ml
    LOVI385132
    Log ind for tilgængelighed
    1 * 1 Items - 112,00 kr