Microplate washers, AquaMax®
Om denne vare
AquaMax® microplate washers are configurable with user interchangeable heads for both 96- and 384-well microplate washing applications, including plate or cell washing.
- Wash heads can be changed in 30 seconds with no tools using innovative head latch mechanism
- All replacement heads are recognised by the washer base, no recalibration required
- Simultaneous well washing without plate indexing or quadrant processing
- Completely programmable via touch screen interface
- No external pumps or computer
- Comprehensive, automated cleaning utilities
- Robotics-friendly design
AquaMax® washers have several important automatic maintenance protocols, since the lack of proper washer maintenance is the number one cause of washer failure. One-Touch AutoClean completely washes system with AquaMax® sterilant; rinses with water and then air dries both the aspiration and dispensing channel to prevent contaminant build up. Auto-shutdown is user-definable and can automatically rinse and purge the AquaMax® washer with air, providing walk away flexibility at the end of the batch.
: AquaMax® washer is available in two configurations. The AquaMax® 2000 has two fluid inlets; the AquaMax® 4000 has four fluid inlets and additional programming capabilities. Both systems have the same onboard touch screen computer for all programming, operation, and maintenance of the system. No external computer is necessary.
2 tilgængelige muligheder nedenfor
Product Details & Documents
AquaMax® microplate washers are configurable with user interchangeable heads for both 96- and 384-well microplate washing applications, including plate or cell washing.
- Wash heads can be changed in 30 seconds with no tools using innovative head latch mechanism
- All replacement heads are recognised by the washer base, no recalibration required
- Simultaneous well washing without plate indexing or quadrant processing
- Completely programmable via touch screen interface
- No external pumps or computer
- Comprehensive, automated cleaning utilities
- Robotics-friendly design
AquaMax® washers have several important automatic maintenance protocols, since the lack of proper washer maintenance is the number one cause of washer failure. One-Touch AutoClean completely washes system with AquaMax® sterilant; rinses with water and then air dries both the aspiration and dispensing channel to prevent contaminant build up. Auto-shutdown is user-definable and can automatically rinse and purge the AquaMax® washer with air, providing walk away flexibility at the end of the batch.
: AquaMax® washer is available in two configurations. The AquaMax® 2000 has two fluid inlets; the AquaMax® 4000 has four fluid inlets and additional programming capabilities. Both systems have the same onboard touch screen computer for all programming, operation, and maintenance of the system. No external computer is necessary.
Specifications for Product Family
Specifikationer
- Catalog No:
- 735-0382
- 735-0383
- Model:
- AquaMax®
- AquaMax®
- Pladetyper:
- 96- and 384-well
- 96- and 384-well
- Dispenseringsvolumenområde:
- 10 - 400 µl
- 10 - 400 µl
- Interface:
- USB and proprietary StakMax connect
- USB and proprietary StakMax connect
Specifikationer
Product Family Options
Produktinformation
- TilgængelighedPrice
- 735-0382AquaMax® 2K includes AquaMax® 2000 microplate washer, 96-plate wash head, 2 assay bottles with level sensor, and 10 l waste bottle with sensorAquaMax® 2K includes AquaMax® 2000 microplate washer, 96-plate wash head, 2 assay bottles with level sensor, and 10 l waste bottle with sensorLog ind for tilgængelighed1 * 1 ItemsSpecifikationer:
More
Specifications
Cat. No.735-0382ModelAquaMax®Pladetyper96- and 384-wellDispenseringsvolumenområde10 - 400 µlInterfaceUSB and proprietary StakMax connect - 735-0383AquaMax® 4K includes AquaMax® 4000 microplate washer, 96-plate wash head, 4 assay bottles with level sensor, and 10 l waste bottle with sensorAquaMax® 4K includes AquaMax® 4000 microplate washer, 96-plate wash head, 4 assay bottles with level sensor, and 10 l waste bottle with sensorLog ind for tilgængelighed1 * 1 ItemsSpecifikationer:
More
Specifications
Cat. No.735-0383ModelAquaMax®Pladetyper96- and 384-wellDispenseringsvolumenområde10 - 400 µlInterfaceUSB and proprietary StakMax connect
Complementary Products
Produktinformation
- Katalog #BeskrivelseAnt. brøndeSterilFarveBrøndvolumenOpløsningModelTilgængelighedPrice
- MLDVPARADIGMSpectraMax® Paradigm®, microplate reader, multi-modeLog ind for tilgængelighed1 * 1 Items - 0,00 kr
- 735-0376SpectraMax® i3x multi-mode detection platformLog ind for tilgængelighed1 * 1 Items - 0,00 kr
- 735-0391SpectraMax® iD3 multi-mode microplate readerLog ind for tilgængelighed1 * 1 Items - 0,00 kr
- 735-0358SpectraMax® iD5 multi-mode microplate reader, with 20×20 well scanningLog ind for tilgængelighed1 * 1 Items - 0,00 kr
Accessories
Produktinformation
- Katalog #TilgængelighedPrice
- Cell wash head, 96-well
Produktinformation
- Katalog #TilgængelighedPrice
- Cell wash head, 384-well
Produktinformation
- Katalog #TilgængelighedPrice
- Bottle with level sensor, 4 l
Produktinformation
- Katalog #TilgængelighedPrice
- Bottle without level sensor, 4 l