Orders LinkContactUsLinkComponent
Blocking buffer, fish gelatin
 :   Biotium
Blocking buffer, fish gelatin
 :  Biotium
Blocking buffer, fish gelatin
Fish gelatin is commonly used in blocking solutions to minimise background noise in immunodetection procedures such as Western Blotting and immunostaining. Fish gelatin can be used to block non-specific binding sites on positively charged nylon or PVDF membranes or for blocking and dilution of antibodies during immunostaining procedures. Unlike BSA or milk, fish gelatin does not contain IgG or serum proteins that could cross-react with mammalian antibodies.
Product Family Options
  • Beskrivelse
    Pakn
  • Fish gelatin blocking buffer, 10% solution
    100 mL
    1 * 100 mL
    1 * 100 mL - 653,00 kr
    653,00 kr
    1 * 100 mL
       653,00 kr
  • Fish gelatin powder, 2×50 µg
    100 g
    1 * 100 g
    1 * 100 g - 517,00 kr
    517,00 kr
    1 * 100 g
       517,00 kr