Crucible, quartz
30 ml, 43×57 mm
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Fused quartz crucible and lid.
- High chemical purity
- High corrosion resistance
- High melting point
- Extreme hardness and low coefficient of thermal expansion
- Excellent electrical insulation qualities and optical transmission from ultra-violet to infra-red
Documents
Specifikationer
- Diameter×H:43×57 mm
- Kapacitet:30 ml