Orders LinkContactUsLinkComponent
Accessories for microcarriers
   Sustainable Options Available
Supplier:   Corning
Accessories for microcarriers
Supplier:  Corning
   Sustainable Options Available
Certificates
Accessories for microcarriers
4 Options Available Below
Product Details & Documents
These polypropylene aseptic transfer screw caps prevents adventitious contamination in for microcarriers bottles during bioreactor loading.
  • Chemically resistant and heat sealable flexible tubing
  • Tubing length with connector 30" (76,2 cm)
  • Disposable and sterile (10⁻⁶ SAL)
Packaging: These caps are individually double bagged.
Caution: *Produktet/produkterne på denne side er uden for anvendelsesområdet for EU -direktiv 2019/904
Specifications for Product Family
Specifications
  • Catalog No:
  • 734-2919
  • 734-2920
  • 734-2921
  • 734-2922
  • Environmentally Preferable:
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Miljøcertificering:
  • RECs, ISO 14001 - Kennebunk
  • RECs, ISO 14001 - Kennebunk
  • RECs, ISO 14001 - Kennebunk
  • RECs, ISO 14001 - Kennebunk
Specifications
Product Family Options
Product Information
  • Beskrivelse
    Availability
    Price
  • Aseptic transfer cap, PP, orange, disposable ,45 mm, male luer lock, for 100 g microcarrier bottle
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.870,00 kr
    1.870,00 kr
    1 * 1 Items
       1.870,00 kr
    Specifications:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    734-2919
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    RECs, ISO 14001 - Kennebunk
      Lav fremstillingspåvirkning
  • Aseptic transfer cap, PP, orange, disposable ,45 mm, MPC, for 100 g microcarrier bottle
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 2.000,00 kr
    2.000,00 kr
    1 * 1 Items
       2.000,00 kr
    Specifications:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    734-2920
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    RECs, ISO 14001 - Kennebunk
      Lav fremstillingspåvirkning
  • Aseptic transfer cap, PP, orange, disposable ,48 mm, male luer lock, for 500 g microcarrier bottle
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.770,00 kr
    1.770,00 kr
    1 * 1 Items
       1.770,00 kr
    Specifications:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    734-2921
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    RECs, ISO 14001 - Kennebunk
      Lav fremstillingspåvirkning
  • Aseptic transfer cap, PP, orange, disposable ,48 mm, MPC, for 500 g microcarrier bottle
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.890,00 kr
    1.890,00 kr
    1 * 1 Items
       1.890,00 kr
    Specifications:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    734-2922
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    RECs, ISO 14001 - Kennebunk