Orders LinkContactUsLinkComponent
Drum fixed angle rotor, DRM 6,14
Drum fixed angle rotor, DRM 6,14
Drum rotor DRM 6.14, 90°, 6 racks, 14300 min-1
Catalog # 521-0294
Supplier:   JOUAN
Drum fixed angle rotor, DRM 6,14
Catalog # 521-0294
Supplier:  JOUAN
Drum fixed angle rotor, DRM 6,14
Drum fixed angle rotor, DRM 6,14
Drum rotor DRM 6.14, 90°, 6 racks, 14300 min-1
Price
14.050,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Drum rotor for use with Thermo Scientific™ Sorvall™ Legend™ 23R and MR23i centrifuges.
Specifications
  • Beskrivelse:
    Drum rotor DRM 6.14, 90°, 6 racks, 14300 min-1