Orders LinkContactUsLinkComponent
Viscosity standards, general purpose
Viscosity standards, general purpose
Leverandør:   CANNON INSTRUMENT
Viscosity standards, general purpose
Leverandør:  CANNON INSTRUMENT
Certifikater
Viscosity standards, general purpose
1 tilgængelige muligheder nedenfor
Product Details & Documents
These certified standards are useful for calibration and verification procedures.
  • It is suitable for all types of viscometers
  • Viscosity data quoted at 20 °C
  • It also helps ensures the reliability of measurements
Certifications: Meets the precision specifications of ASTM D 445/446 and ISO 3104/3105, and are traceable to the NIST.
Product Family Options
Produktinformation
  • Antal
    Viskositet
    Pakning
    Pakn
    Tilgængelighed
    Price
  • S6
    10 mm²/s
    Glass bottle
    500 mL
    Discontinued
  • S600
    1500 mm²/s
    Glass bottle
    500 mL
    Discontinued
  • S8000
    33000 mm²/s
    500 mL
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 500 mL
    1 * 500 mL
    Specifikationer:
    Marketsource
    Marketsource
  • S30000
    Glass bottle
    1
    Discontinued