Dicalite® speed plus diatomaceous filter-aid
Plastic bottle, 1 kg
Leverandør: Thermo Fisher Scientific
Dicalite® speed plus diatomaceous filter-aid
Katalog # ACRO123380010
Leverandør: Thermo Fisher Scientific
CAS nummer: 68855-54-9
Gå til:
Product Details & Documents
- CAS nummer:68855-54-9
- MDL nummer:MFCD00132803
- Opbevaringstemperatur:Stue temperatur
Documents
Specifikationer
Specifikationstestresultater
- Appearance:White to off-white Powder
- Additional info:typical properties
- Ratio:85 to 115 Permeability flow ratio
Specifikationer
- Pakning:Plastic bottle
- Pakn:1 kg