Orders LinkContactUsLinkComponent
X-β-D-cellobioside
X-β-D-cellobioside
25 mg
Katalog # 10026.
Irriterende
Leverandør:   Biotium
X-β-D-cellobioside
Katalog # 10026.
Leverandør:  Biotium
CAS nummer:  177966-52-8
X-β-D-cellobioside
X-β-D-cellobioside
25 mg
CAS nummer:  177966-52-8
Price
1.780,00 kr
Log ind for tilgængelighed
1 * 25 mg
Product Details & Documents
X-β-D-cellobioside is a chromogenic substrate for β-cellobiosidase.
  • Cellobiosidase Substrate
White solid soluble in DMSO.
Enzymatic cleavage of the substrate generates a dark blue-coloured precipitate.
Caution: Store at −20 °C and protect from light.
Synonymer:  5-Bromo-4-chloro-3-indolyl-β-D-cellobioside
  • CAS nummer:177966-52-8
  • Formel:C₂₀H₂₅BrClNO₁₁
  • Molecular Weight:570.78 g/mol
  • Opbevaringstemperatur:Fryser
Specifikationer
Specifikationer
  • Pakn:
    25 mg