Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Phenol saturated, Pierce, Pierce™

Phenol saturated, Pierce, Pierce™

400 ml
Katalog # PIER17914
Leverandør:   Thermo Fisher Scientific
Phenol saturated, Pierce, Pierce™
Katalog # PIER17914
CAS nummer:  108-95-2
Phenol saturated, Pierce, Pierce™
Phenol saturated, Pierce, Pierce™

400 ml

CAS nummer:  108-95-2
Price
1.960,00 kr
Log ind for tilgængelighed
1 * 400 mL
Product Details & Documents

Saturated phenol is a phenol supplied as a saturated solution and accompanied by a bottle of buffer to adjust the pH for typical molecular biology applications.

  • pH 6,6 or pH 7,9 for use in typical DNA purifications
  • Prepared from high purity phenol, double-distilled under nitrogen
  • Phenol is saturated completely and extra buffer is added as an overlay to prevent oxidation
Synonymer:  Phenylalcohol, Carbolic acid, Hydroxybenzene
  • CAS nummer:108-95-2
  • Formel:C₆H₅OH
  • MDL nummer:MFCD00002143
  • Molecular Weight:94.11 g/mol
  • FN:1671
  • ADR:6.1,II
Specifikationer

Specifikationer

  • Pakn:
    400 ml

Specifikationer