ESI negative ion calibration solution, LTQ, Pierce™
Pierce™ negative ion calibration solution, LTQ ESI Pierce™, 10 mL
Leverandør: Thermo Fisher Scientific
ESI negative ion calibration solution, LTQ, Pierce™
Katalog # PIER88324
Leverandør: Thermo Fisher Scientific
CAS nummer:
Gå til:
Product Details & Documents
Ion calibration solution are mixtures of highly purified ionizable molecules specifically designed for negative mode calibration of ion trap and orbitrap instruments.
- Mass spectrometry grade reagents in a non-leachable container
- Load the mass reference standard into a syringe and inject into the instrument
- Strong peaks—mixture of SDS, sodium taurocholate and Ultramark 1621 in a acetonitrile/methanol/acetic solution.
It is a ready-to-use liquid formulation ideal for quickly performing the routine calibration required to maintain the robust performance of Thermo Scientific mass spectrometers.
Documents
Certifikater
Søg efter partinummerSpecifikationer
Specifikationer
- Pakn:10 mL
- Beskrivelse:Pierce™ negative ion calibration solution, LTQ ESI Pierce™