Orders LinkContactUsLinkComponent
Test kits, formaldehyde, Spectroquant®, Supelco®
Test kits, formaldehyde, Spectroquant®, Supelco®
Supplier:  Merck
Certificates
Test kits, formaldehyde, Spectroquant®, Supelco®
2 Options Available Below
Product Details & Documents
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request). Spectroquant® cell test kits.
  • Very user friendly, validated methods
  • All the necessary reagents for the analysis are contained in the cells for most the test parameters
  • Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
  • The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times
Spectroquant® reagent test kits
- Highly stable, ready to use reagent mixtures in a user friendly design
- Free selection of appropriate cell format according to the measuring range
- Long shelf life up to 3 years - all at room temperature
- Automatic selection of the correct method with help of AutoSelector barcode identification
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.
Specifications for Product Family
Specifications
Specifications
  • Catalog No:
  • 1.14500.0001
  • 1.14678.0001
  • M-Clarity quality level:
  • MQ100
  • MQ100
Specifications
Product Family Options
Product Information
  • Beskrivelse
    Område
    Kemisk metode
    Til enhed
    Pakn
    Availability
    Price
  • Formaldehydkuvettetest
    0,10 - 8,00 mg/l HCHO
    Chromotropic acid
    Prove, Nova 30/60
    25 Tests
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each - 1.110,00 kr
    1.110,00 kr
    1 * 1 Each
       1.110,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    1.14500.0001
    M-Clarity quality level
    MQ100
  • Formaldehydtest
    0,02 - 8,00 mg/l HCHO
    Chromotropic acid
    Prove, Nova 60
    100 Tests
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each - 1.810,00 kr
    1.810,00 kr
    1 * 1 Each
       1.810,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    1.14678.0001
    M-Clarity quality level
    MQ100