Om denne vare
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request). Spectroquant® cell test kits.
- Very user friendly, validated methods
- All the necessary reagents for the analysis are contained in the cells for most the test parameters
- Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
- The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times
Spectroquant® reagent test kits
- Highly stable, ready to use reagent mixtures in a user friendly design
- Free selection of appropriate cell format according to the measuring range
- Long shelf life up to 3 years - all at room temperature
- Automatic selection of the correct method with help of AutoSelector barcode identification
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.
Note: For determination of the total content of lead please use one of the Crack Sets (10 or 10C) before the photometric procedure.
2 tilgængelige muligheder nedenfor
Product Details & Documents
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request). Spectroquant® cell test kits.
- Very user friendly, validated methods
- All the necessary reagents for the analysis are contained in the cells for most the test parameters
- Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
- The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times
Spectroquant® reagent test kits
- Highly stable, ready to use reagent mixtures in a user friendly design
- Free selection of appropriate cell format according to the measuring range
- Long shelf life up to 3 years - all at room temperature
- Automatic selection of the correct method with help of AutoSelector barcode identification
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.
Note: For determination of the total content of lead please use one of the Crack Sets (10 or 10C) before the photometric procedure.
Documents
Specifications for Product Family
Specifikationer
Specifikationer
- Catalog No:
- 1.09717.0001
- 1.14833.0001
- M-Clarity quality level:
- MQ100
- MQ100
Specifikationer
Product Family Options
Produktinformation
- OmrådeKemisk metodeTil enhedPaknTilgængelighedPrice
- 1.09717.0001BlytestBlytest0,010 - 5,00 mg/l PbPARProve, Nova 60, Multy, Move50 TestsLog ind for tilgængelighed1 * 1 Each1.200,00 krSpecifikationer:
More
Specifications
Cat. No.1.09717.0001M-Clarity quality levelMQ100 - 1.14833.0001BlykuvettetestBlykuvettetest0,10 - 5,00 mg/l PbPARProve, Nova 30/60, Multy, Move25 TestsLog ind for tilgængelighed1 * 1 Each1.190,00 krSpecifikationer:
More
Specifications
Cat. No.1.14833.0001M-Clarity quality levelMQ100