Testkits, chlordioxidtest til Spectroquant®, Supelco®
Chlordioxidtest, 0,020 - 10,00 mg/l ClO₂, DPD, Prove, Nova 60, Multy, Move, Picco, 200 Tests
Leverandør: Merck
Testkits, chlordioxidtest til Spectroquant®, Supelco®
Katalog # 1.00608.0001
Leverandør: Merck
CAS nummer:
Gå til:
Product Details & Documents
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request).
- Highly stable, ready to use reagent mixtures in a user friendly design
- Free selection of appropriate cell format according to the measuring range
- Long shelf life up to 3 years - all at room temperature
- Automatic selection of the correct method with help of AutoSelector barcode identification
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.
Documents
Certifikater
Søg efter partinummerHow to use Testkits?
Videoer
Merck Segment Brochure Merck Spectroquant® Move DC Mobile Colorimeter for Disinfection Control
Produktlitteratur
msds_100608
SDS
Specifikationer
Specifikationer
- Kemisk metode:DPD
- Pakn:200 Tests
- Beskrivelse:Chlordioxidtest
- Til enhed:Prove, Nova 60, Multy, Move, Picco
- Område:0,020 - 10,00 mg/l ClO₂
- Cat. No.:1.00608.0001
- M-Clarity quality level:MQ100