Orders LinkContactUsLinkComponent
IgM purification kit , Pierce™
IgM purification kit , Pierce™
Leverandør:   Thermo Fisher Scientific
IgM purification kit , Pierce™
Certifikater
IgM purification kit , Pierce™
IgM purification kit , Pierce™
3 tilgængelige muligheder nedenfor
Product Details & Documents
The Pierce™ IgM Purification Kit is an easy to use affinity matrix and buffer system to purify IgM from mouse ascites fluid. The method uses immobilised mannan binding protein (MBP), which binds to IgM in the kit-supplied buffers.
  • Immobilised MBP allows specific purification of mouse IgM from ascites fluid
  • Non denaturing, near-neutral buffer conditions ensure recovery of functional antibodies
  • Stable ligand and resins can be regenerated and reused at least ten times
Delivery information: Kit includes MBP Agarose column, 5 m, 1 column; IgM Binding buffer, 800 ml; IgM Elution buffer, 500 ml; MBP Column Preparation buffer, 50 ml; and Column extender.
Product Family Options
Produktinformation
  • Pakn
    Tilgængelighed
    Price
  • PIER44897
    Pierce™ IgM Purification kit, sufficient for 10 IgM purifications, each from 0,5 ml mouse ascites samples
    Pierce™ IgM Purification kit, sufficient for 10 IgM purifications, each from 0,5 ml mouse ascites samples
    1 KIT
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 9.350,00 kr
    9.350,00 kr
    1 * 1 Each
       9.350,00 kr
  • PIER21018
    MBP Column preparation buffer
    MBP Column preparation buffer
    50 ml
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 50 mL
    1 * 50 mL - 552,00 kr
    552,00 kr
    1 * 50 mL
       552,00 kr
  • PIER22212
    MBP Agarose, mannan binding protein on 4% beaded agarose
    MBP Agarose, mannan binding protein on 4% beaded agarose
    10 mL
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 10 mL
    1 * 10 mL - 13.810,00 kr
    13.810,00 kr
    1 * 10 mL
       13.810,00 kr