Orders LinkContactUsLinkComponent
North2South™ Hybridisation Buffer
North2South™ Hybridisation Buffer
North2South™ hybridisation buffer, pH 6,9, 125 ml
Katalog # PIER37549
Leverandør:   Thermo Fisher Scientific
North2South™ Hybridisation Buffer
Katalog # PIER37549
CAS nummer:  
North2South™ Hybridisation Buffer
North2South™ Hybridisation Buffer
North2South™ hybridisation buffer, pH 6,9, 125 ml
CAS nummer:  
Price
1.920,00 kr
Log ind for tilgængelighed
1 * 125 mL
Product Details & Documents
North2South™ hybridisation buffer is an aqueous buffer which is used with the North2South™ chemiluminescent detection kit.
Specifikationer
Specifikationer
  • Pakn:
    125 ml
  • Beskrivelse:
    North2South™ hybridisation buffer, pH 6,9