PageBlue™ protein staining solution
PageBlue™ Protein Staining Solution, 1 L
Leverandør: Thermo Fisher Scientific
PageBlue™ protein staining solution
Katalog # PIER24620
Leverandør: Thermo Fisher Scientific
CAS nummer:
Gå til:
Product Details & Documents
PageBlue Protein Staining Solution is a sensitive, economical formulation of colloidal coomassie G-250 dye for endpoint staining of proteins in polyacrylamide gels and on PVDF membranes without methanol or acetic acid.
- Easy-to-use—simply soak the gel in the ready-to-use solution and observe stained bands without destaining
- Staining time—25 to 40 minute protocol
- Safe—does not contain methanol or acetic acid
- Dynamic range—linear dynamic range of 5 to 500 ng
- Economical—can be reused up to three times
The simple PageBlue Protein Staining Solution protocol is sensitive, efficient, and eliminates the concern of over-staining gels, even after overnight staining. This protein stain delivers a dynamic range of 5 to 500 ng, which is approximately ten times more sensitive than traditional coomassie R-250-based dyes.
: For Research Use Only. Not for use in diagnostic procedures. Upon receipt store product at room temperature. Product shipped at ambient temperature
Certifikater
Søg efter partinummerSpecifikationer
Specifikationer
- Pakn:1 L
- Beskrivelse:PageBlue™ Protein Staining Solution