Orders LinkContactUsLinkComponent
ISE sample conditioning buffers and solutions
ISE sample conditioning buffers and solutions
 :   WTW
ISE sample conditioning buffers and solutions
 :  WTW
ISE sample conditioning buffers and solutions
Ion strength adjustment solutions have multiple tasks: TISAB for fluoride measurements does not only adjust the pH value, by also decomplexes the fluoride-containing complex compounds. Other ion strength adjustment solutions ensure an adapted pH value as well as the addition of a suitable conducting salt to correctly capture the concentration.
Product Family Options
  • Beskrivelse
    pH
    Pakning
    Pakn
  • ISA/K, ionic strength adjustment solution for potassium ions
    Bottle
    250 mL
    1 * 250 mL
    1 * 250 mL - 1.570,00 kr
    1.570,00 kr
    1 * 250 mL
       1.570,00 kr
  • TISAB buffer-opløsning (F800)
    Bottle
    4
    1 * 4 Items
    1 * 4 Items - 3.130,00 kr
    3.130,00 kr
    1 * 4 Items
       3.130,00 kr
  • ISA/FK-opløsning, NaNO₃, 5 mol/L (Cl800/Cu800)
    Bottle
    500 mL
    1 * 500 mL
    1 * 500 mL - 1.570,00 kr
    1.570,00 kr
    1 * 500 mL
       1.570,00 kr
  • ISA/Ca, ionic strength adjustment solution for calcium ions
    Bottle
    250 mL
    1 * 250 mL
    1 * 250 mL - 1.570,00 kr
    1.570,00 kr
    1 * 250 mL
       1.570,00 kr
  • TISAB/NO₃, total ionic strength adjustment buffer for nitrate
    Bottle
    4
    1 * 4 Items
    1 * 4 Items - 3.420,00 kr
    3.420,00 kr
    1 * 4 Items
       3.420,00 kr
  • ISA/Na, ionic strength adjustment solution for sodium
    11
    Bottle
    1 L
    1 * 1 L
    1 * 1 L - 1.260,00 kr
    1.260,00 kr
    1 * 1 L
       1.260,00 kr