Orders LinkContactUsLinkComponent
Hybridisation buffer, SSPE 20X, AccuGENE™
Hybridisation buffer, SSPE 20X, AccuGENE™
AccuGENE™ 20X SSPE buffer, 3,0 M NaCl, 0,2 M NaH₂PO₄, H₂O, 0,02 M EDTA, pH 7,4, 1 L
Katalog # 733-1638
Leverandør:   LONZA
Hybridisation buffer, SSPE 20X, AccuGENE™
Katalog # 733-1638
Leverandør:  LONZA
CAS nummer:  
Hybridisation buffer, SSPE 20X, AccuGENE™
AccuGENE™ 20X SSPE buffer, 3,0 M NaCl, 0,2 M NaH₂PO₄, H₂O, 0,02 M EDTA, pH 7,4, 1 L
CAS nummer:  
Price
717,00 kr
Log ind for tilgængelighed
1 * 1 L
Product Details & Documents
AccuGENE™ electrophoresis buffers are ready-to-use solutions ideal for a wide range of molecular biology applications.
  • Ready to use solutions
  • Reliable manufactured according to strict quality control standards to ensure lot to lot consistency
  • Guaranteed DNase-, RNase- and protease-free
Each reagent is prepared with 18 megOhm water, filtered using a 0,2 micron filter, and filled into sterile bottles.
Documents
Specifikationer
Specifikationer
  • Pakn:
    1 L
  • Beskrivelse:
    AccuGENE™ 20X SSPE buffer, 3,0 M NaCl, 0,2 M NaH₂PO₄, H₂O, 0,02 M EDTA, pH 7,4
Specifikationer