β-Caryophyllene, liquid
Leverandør: MP Biomedicals
Gå til:
Om denne vare
(-)-trans-Caryophyllene is a natural bicyclic sesquiterpene that is a constituent of many essential oils.
Synonymer: (1R,4E,9S)-4,11,11-trimethyl-8-methylene-bicyclo[7.2.0]undec-4-ene, (1R,9S,E)-4,11,11-Trimethyl-8-methylenebicyclo[7.2.0]undec-4-ene, (−)-β-Caryophyllene, (-)-trans-caryophyllene, NSC 11906
- CAS nummer:87-44-5
- Formel:C₁₅H₂₄
- MDL nummer:MFCD00075925
- Molecular Weight:204.36 g/mol
2 tilgængelige muligheder nedenfor
Product Details & Documents
(-)-trans-Caryophyllene is a natural bicyclic sesquiterpene that is a constituent of many essential oils.
Synonymer: (1R,4E,9S)-4,11,11-trimethyl-8-methylene-bicyclo[7.2.0]undec-4-ene, (1R,9S,E)-4,11,11-Trimethyl-8-methylenebicyclo[7.2.0]undec-4-ene, (−)-β-Caryophyllene, (-)-trans-caryophyllene, NSC 11906
- CAS nummer:87-44-5
- Formel:C₁₅H₂₄
- MDL nummer:MFCD00075925
- Molecular Weight:204.36 g/mol